Gene Transcripts

Gene Symbol
Tmem14a
Entrez Gene ID
75712
Gene Name
transmembrane protein 14A
Gene Aliases
5730496E24Rik, C85783, PTD011
Chromosome Location
Chr. 1: 21166436 - 21230167 on Build GRCm38
UniGene ID
Mm.304763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290679.1 NP_001277608.1 4, 5 592
GenBank mRNA AK017734.1 - 4, 5 507

View Full Product Details