Gene Transcripts

Gene Symbol
Spink12
Entrez Gene ID
78242
Gene Name
serine peptidase inhibitor, Kazal type 12
Gene Aliases
9230117E20Rik
Chromosome Location
Chr. 18: 44104523 - 44108543 on Build GRCm38
UniGene ID
Mm.99782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030061.3 NP_084337.1 1 209
GenBank mRNA AK020352.1 - 1 110
AK136616.1 - 1 210
BC145914.1 - 1 95 40131217
BC145916.1 - 1 95 40131219
DQ437332.1 - 1 268

View Full Product Details