Gene Transcripts

Gene Symbol
Gpr87
Entrez Gene ID
84111
Gene Name
G protein-coupled receptor 87
Gene Aliases
Chromosome Location
Chr. 3: 59178904 - 59195468 on Build GRCm38
UniGene ID
Mm.60267
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF295366.1 - 1 95
AK080394.1 - 3 433
BC125653.1 - 1 124 40130168
BC125655.1 - 1 124 40130170

View Full Product Details