Gene Transcripts

Gene Symbol
Fancg
Entrez Gene ID
60534
Gene Name
Fanconi anemia, complementation group G
Gene Aliases
AU041407, Xrcc9
Chromosome Location
Chr. 4: 43002337 - 43010568 on Build GRCm38
UniGene ID
Mm.23122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163233.1 NP_001156705.1 3 485
NM_053081.2 NP_444311.1 3 485
GenBank mRNA AF112439.1 - 3 294
AY049715.1 - 3 285
BC050890.1 - 4 589
BC139763.1 - 3 288 40110334
BC141282.1 - 3 301 9055894

View Full Product Details