Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164311.1 | NP_001157783.1 | 12 | 2033 | |
NM_053083.3 | NP_444313.3 | 12 | 2030 | ||
GenBank mRNA | AF338440.1 | - | 11 | 1925 | |
AK014331.1 | - | 12 | 2033 | ||
BC068307.1 | - | 12 | 1972 | 30059745 |