Gene Transcripts

Gene Symbol
Csmd1
Entrez Gene ID
94109
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
B930082H09, mKIAA1890
Chromosome Location
Chr. 8: 15892542 - 17535385 on Build GRCm38
UniGene ID
Mm.436988
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053171.2 NP_444401.2 2 625
GenBank mRNA AK038679.1 - 2 606
AK048698.1 - 2 608
AK048713.1 - 2 827
AK134585.1 - 2 626
AY017475.1 - 2 522

View Full Product Details