Gene Transcripts
- Gene Symbol
- Optc
- Entrez Gene ID
- 269120
- Gene Name
- opticin
- Gene Aliases
- Chromosome Location
- Chr. 1: 133897193 - 133909714 on Build GRCm38
- UniGene ID
- Mm.59985
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160420.2 |
NP_001153892.1
|
3, 4 |
420 |
|
NM_001160421.2 |
NP_001153893.1
|
2, 3 |
531 |
|
NM_001160422.1 |
NP_001153894.1
|
2, 3 |
531 |
|
NM_054076.3 |
NP_473417.2
|
2, 3 |
531 |
|
GenBank mRNA |
AF333980.1 |
- |
2, 3 |
530 |
|
AK053318.1 |
- |
2, 3 |
314 |
|
AK153750.1 |
- |
3, 4 |
421 |
|
AY077682.1 |
- |
2, 3 |
523 |
|
BC099435.1 |
- |
2, 3 |
364 |
30788813 |
BC115928.1 |
- |
2, 3 |
484 |
40104738 |
View Full Product Details