Gene Transcripts

Gene Symbol
Ppap2b
Entrez Gene ID
67916
Gene Name
phosphatidic acid phosphatase type 2B
Gene Aliases
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b
Chromosome Location
Chr. 4: 105157347 - 105232767 on Build GRCm38
UniGene ID
Mm.348326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080555.2 NP_542122.1 6 1400
GenBank mRNA AK011276.1 - 6 1398
AK076183.1 - 6 1096
AK159136.1 - 6 1486
AK160056.1 - 6 1300
BC005558.1 - 6 1387 3964817

View Full Product Details