Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133716.3 | NP_598477.2 | 10 | 2463 | |
GenBank mRNA | AB029329.1 | - | 1 | 871 | |
AK007666.1 | - | 2 | 1064 | ||
BC031581.1 | - | 10 | 2452 | ||
BC032217.1 | - | 11 | 2308 | ||
BC051481.1 | - | 1 | 365 | ||
BC052413.1 | - | 10 | 2461 | 5720177 |