Gene Transcripts

Gene Symbol
Macrod1
Entrez Gene ID
107227
Gene Name
MACRO domain containing 1
Gene Aliases
AI604841, AW743046, D930010J01Rik, Lrp16
Chromosome Location
Chr. 19: 7056715 - 7198062 on Build GRCm38
UniGene ID
Mm.439977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134147.4 NP_598908.2 3 563
GenBank mRNA AK052998.1 - 3 522
BC008653.1 - 3 288 3597023
BQ126199.1 - 3 563

View Full Product Details