Gene Transcripts

Gene Symbol
Med10
Entrez Gene ID
28077
Gene Name
mediator complex subunit 10
Gene Aliases
AA959813, AI385605, C78613, D13Wsu50e
Chromosome Location
Chr. 13: 69802348 - 69816104 on Build GRCm38
UniGene ID
Mm.491490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138596.2 NP_613062.1 3 317
GenBank mRNA AK003862.1 - 3 311
AK013992.1 - 3 321
AK156498.1 - 3 326
AK215905.1 - 3 165
BC048638.1 - 3 310 6771841
BC056438.1 - 3 306 6818638
BC059231.1 - 2 2799
BC089312.1 - 2 1862

View Full Product Details