Gene Transcripts
- Gene Symbol
- Dppa3
- Entrez Gene ID
- 73708
- Gene Name
- developmental pluripotency-associated 3
- Gene Aliases
- 2410075G02Rik, PCG7, PGC7, Stella
- Chromosome Location
- Chr. 6: 122626424 - 122630271 on Build GRCm38
- UniGene ID
- Mm.27982
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139218.1 |
NP_631964.1
|
3, 4 |
448 |
|
GenBank mRNA |
AB072734.1 |
- |
3, 4 |
349 |
|
AF490347.1 |
- |
3, 4 |
453 |
|
AK010723.1 |
- |
3, 4 |
389 |
|
AK135769.1 |
- |
3, 4 |
453 |
|
AK136263.1 |
- |
3, 4 |
453 |
|
AY082485.1 |
- |
3, 4 |
448 |
|
AY134859.1 |
- |
3, 4 |
460 |
|
BC099433.1 |
- |
3, 4 |
426 |
30933569 |
BC100331.1 |
- |
3, 4 |
423 |
30934920 |
BC107340.1 |
- |
3, 4 |
380 |
40055218 |
BC107341.1 |
- |
3, 4 |
380 |
40055220 |
View Full Product Details