Gene Transcripts

Gene Symbol
Thyn1
Entrez Gene ID
77862
Gene Name
thymocyte nuclear protein 1
Gene Aliases
D730042P09Rik, HSPC144, MDS012, MY105, Thy28
Chromosome Location
Chr. 9: 26999659 - 27007334 on Build GRCm38
UniGene ID
Mm.16925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144543.2 NP_653126.1 3, 4 371
GenBank mRNA AB080370.1 - 2, 3 295
AK077639.1 - 3, 4 341
BC016594.1 - 3, 4 325 2631627
CB236103.1 - 2, 3 564

View Full Product Details