Gene Transcripts

Gene Symbol
Fam49b
Entrez Gene ID
223601
Gene Name
family with sequence similarity 49, member B
Gene Aliases
0910001A06Rik, AW122079
Chromosome Location
Chr. 15: 63929094 - 64060452 on Build GRCm38
UniGene ID
Mm.246108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144846.5 NP_659095.1 4 453
GenBank mRNA AK028062.1 - 4 454
AK075613.1 - 5 398
AK205521.1 - 3 202
BC011343.1 - 4 271 4012217

View Full Product Details