Gene Transcripts

Gene Symbol
Tspan14
Entrez Gene ID
52588
Gene Name
tetraspanin 14
Gene Aliases
AA958793, D14Ertd226e, Tm4sf14
Chromosome Location
Chr. 14: 40906427 - 40966807 on Build GRCm38
UniGene ID
Mm.28876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145928.1 NP_666040.1 8 749
GenBank mRNA AK030593.1 - 8 751
AK156671.1 - 8 762
AK215120.1 - 1 71
BC024611.1 - 8 754 4188261
BC025568.1 - 8 763 5322531
BC026574.1 - 8 749 4954874

View Full Product Details