Gene Transcripts

Gene Symbol
Dmrt2
Entrez Gene ID
226049
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
Terra
Chromosome Location
Chr. 19: 25672391 - 25679013 on Build GRCm38
UniGene ID
Mm.275572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145831.3 NP_665830.1 2 800
GenBank mRNA AF539811.1 - 2 800
AK044891.1 - 2 801
AK140337.1 - 2 792
AK140347.1 - 2 795

View Full Product Details