Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161406.1 | NP_001154878.1 | 14, 15 | 1834 | |
NM_145890.2 | NP_665897.2 | 12, 13 | 1466 | ||
GenBank mRNA | AF411211.1 | - | 12, 13 | 1466 | |
AF411212.1 | - | 14, 15 | 1805 | ||
AK076307.1 | - | 14, 15 | 1820 | ||
AK161048.1 | - | 14, 15 | 1716 | ||
BC013080.1 | - | 14, 15 | 1774 | 3499860 | |
BC037233.1 | - | 14, 15 | 1826 | 4951512 |