Gene Transcripts

Gene Symbol
Emc1
Entrez Gene ID
230866
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
2700016F22Rik
Chromosome Location
Chr. 4: 139352587 - 139378735 on Build GRCm38
UniGene ID
Mm.394288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039200.2 NP_001034289.1 23 5325
NM_146157.4 NP_666269.2 24 5334
GenBank mRNA AK049061.1 - 24 5322
AK077496.1 - 24 5326
BC129804.1 - 25 5256 40087024

View Full Product Details