Gene Transcripts

Gene Symbol
NRBP1
Entrez Gene ID
29959
Gene Name
nuclear receptor binding protein 1
Gene Aliases
BCON3, MADM, MUDPNP, NRBP
Chromosome Location
Chr. 2: 27427790 - 27442259 on Build GRCh38
UniGene ID
Hs.515876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321357.1 NP_001308286.1 3, 4 1050
NM_001321358.1 NP_001308287.1 2, 3 423
NM_001321359.1 NP_001308288.1 2, 3 418
NM_001321361.1 NP_001308290.1 2, 3 423
NM_001321362.1 NP_001308291.1 2, 3 423
NM_001321363.1 NP_001308292.1 2, 3 423
NM_013392.3 NP_037524.1 2, 3 418
GenBank mRNA AF113249.1 - 2, 3 330
AK001946.1 - 2, 3 308
AK027538.1 - 2, 3 326
AK122664.1 - 3, 4 1050
AK223136.1 - 2, 3 314
AK225540.1 - 2, 3 344
AK301345.1 - 3, 4 261
AL136682.1 - 2, 3 305
BC001221.1 - 2, 3 332 3534543
BQ949042.1 - 2, 3 364
DA161015.1 - 2, 3 424

View Full Product Details