Gene Transcripts

Gene Symbol
Katnal1
Entrez Gene ID
231912
Gene Name
katanin p60 subunit A-like 1
Gene Aliases
Chromosome Location
Chr. 5: 148871584 - 148929134 on Build GRCm38
UniGene ID
Mm.397405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153572.2 NP_705800.1 2 234
GenBank mRNA AK049620.1 - 2 588
BC030434.1 - 2 221 5369313
BE653455.1 - 2 234

View Full Product Details