Gene Transcripts

Gene Symbol
PDHB
Entrez Gene ID
5162
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Gene Aliases
PDHBD, PDHE1-B, PHE1B
Chromosome Location
Chr. 3: 58427630 - 58433852 on Build GRCh38
UniGene ID
Hs.161357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK225621.1 - 10 1204
J03576.1 - 10 1199
M34056.1 - 9 1109
M34479.1 - 10 1181
M54788.1 - 10 1186
X57778.1 - 10 1200

View Full Product Details