Gene Transcripts

Gene Symbol
Scarf2
Entrez Gene ID
224024
Gene Name
scavenger receptor class F, member 2
Gene Aliases
AI194363, NSR1, SREC-II, SRECRP-1, Scarf1, Scatf1, Srec2, SrecII, W53987, W65560
Chromosome Location
Chr. 16: 17797282 - 17808294 on Build GRCm38
UniGene ID
Mm.194950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153790.2 NP_722485.1 3 325
GenBank mRNA AB024432.1 - 3 292
AF522197.1 - 3 326
AK142378.1 - 3 293
AK143208.1 - 3 167
BC059226.1 - 3 257
BC060084.1 - 3 257
BC139758.1 - 3 278 40047931

View Full Product Details