Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163626.1 | NP_001157098.1 | 3 | 597 | |
NM_172204.4 | NP_757341.3 | 3 | 597 | ||
GenBank mRNA | AF539798.1 | - | 3 | 316 | |
AK087263.1 | - | 3 | 523 | ||
AY255770.1 | - | 3 | 316 | ||
BC047532.1 | - | 3 | 588 | 4988389 |