Gene Transcripts

Gene Symbol
Phyhd1
Entrez Gene ID
227696
Gene Name
phytanoyl-CoA dioxygenase domain containing 1
Gene Aliases
Chromosome Location
Chr. 2: 30266203 - 30282149 on Build GRCm38
UniGene ID
Mm.297949
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252568.2 NP_001239497.1 11 1540
NM_001252570.1 NP_001239499.1 11 1230
NM_001252571.1 NP_001239500.1 10 1167
NM_001281829.1 NP_001268758.1 12 1256
NM_172267.4 NP_758471.1 12 1609
GenBank mRNA AK005293.1 - 12 1610
AK138607.1 - 12 1284
BC039982.1 - 12 1251
BU701903.1 - 2 392

View Full Product Details