Gene Transcripts

Gene Symbol
Vezt
Entrez Gene ID
215008
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
A630071D13Rik, AI848282
Chromosome Location
Chr. 10: 93961522 - 94035799 on Build GRCm38
UniGene ID
Mm.124177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK031848.1 - 4 488
AK036017.1 - 4 436
AK041354.1 - 4 465
AK042540.1 - 4 430
AY753561.1 - 4 392
AY753562.1 - 4 392
BC056428.1 - 4 423 5716245
DQ025533.1 - 4 392

View Full Product Details