Gene Transcripts

Gene Symbol
EIF2AK1
Entrez Gene ID
27102
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases
HCR, HRI
Chromosome Location
Chr. 7: 6022247 - 6059229 on Build GRCh38
UniGene ID
Hs.520205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134335.1 NP_001127807.1 3 540
NM_014413.3 NP_055228.2 3 540
GenBank mRNA AB037790.1 - 3 465
AF100784.1 - 3 429
AF147094.1 - 3 475
AF183414.1 - 3 427
AF255050.1 - 3 485
AK075192.1 - 3 468
AK290327.1 - 3 473
AL136563.1 - 3 445
AL834494.1 - 3 445
BC006524.2 - 3 472 2989989
DC341983.1 - 3 540

View Full Product Details