Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172711.2 | NP_766299.1 | 1, 2 | 232 | |
GenBank mRNA | AK047411.1 | - | 1, 2 | 101 | |
AK053420.1 | - | 1, 2 | 206 | ||
AK084627.1 | - | 1, 2 | 213 | ||
AK140534.1 | - | 1, 2 | 191 | ||
AK160956.1 | - | 1, 2 | 251 | ||
AK165012.1 | - | 1, 2 | 221 | ||
BC057962.1 | - | 1, 2 | 186 | 5353891 | |
BY743247.1 | - | 1, 2 | 233 |