Gene Transcripts
- Gene Symbol
- Dok7
- Entrez Gene ID
- 231134
- Gene Name
- docking protein 7
- Gene Aliases
- A930013K19Rik, AW049091, Dok-7, EF-12, Oit5
- Chromosome Location
- Chr. 5: 35056566 - 35087839 on Build GRCm38
- UniGene ID
- Mm.205483
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172708.3 |
NP_766296.1
|
6, 7 |
1459 |
|
GenBank mRNA |
AK044445.1 |
- |
6, 7 |
1460 |
|
View Full Product Details