Gene Transcripts

Gene Symbol
Elmo3
Entrez Gene ID
234683
Gene Name
engulfment and cell motility 3
Gene Aliases
9930107J06, BC058752, CED-12, Ced12
Chromosome Location
Chr. 8: 105305399 - 105310623 on Build GRCm38
UniGene ID
Mm.237966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172760.3 NP_766348.3 6, 7 672
GenBank mRNA AK037067.1 - 6, 7 665
AK157875.1 - 610
BC018516.1 - 4, 5 561 3155913
BC026617.1 - 1, 2 617
BC058752.1 - 6, 7 655 6485393
BC138887.1 - 6, 7 629 8861909

View Full Product Details