Gene Transcripts

Gene Symbol
Ttll8
Entrez Gene ID
239591
Gene Name
tubulin tyrosine ligase-like family, member 8
Gene Aliases
1700019P01Rik, 4933401B17
Chromosome Location
Chr. 15: 88889778 - 88955641 on Build GRCm38
UniGene ID
Mm.479911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172818.3 NP_766406.2 3 668
GenBank mRNA AK029745.1 - 3 669
AK030151.1 - 3 661
AM690752.1 - 3 325
BC116293.1 - 3 309 40094952
BC116294.1 - 3 309 40094955

View Full Product Details