Gene Transcripts

Gene Symbol
Slc6a11
Entrez Gene ID
243616
Gene Name
solute carrier family 6 (neurotransmitter transporter, GABA), member 11
Gene Aliases
D930045G19Rik, E130202I16Rik, GAT4, Gabt4, Gat3
Chromosome Location
Chr. 6: 114131241 - 114249886 on Build GRCm38
UniGene ID
Mm.44683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172890.3 NP_766478.1 2 397
GenBank mRNA AK053078.1 - 2 398
AK053261.1 - 2 239
AK140423.1 - 2 400

View Full Product Details