Gene Transcripts

Gene Symbol
Dcun1d3
Entrez Gene ID
233805
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae)
Gene Aliases
1700020A13Rik, BC030335
Chromosome Location
Chr. 7: 119853158 - 119896169 on Build GRCm38
UniGene ID
Mm.31539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163703.1 NP_001157175.1 2 570
NM_173408.3 NP_775584.1 2 589
GenBank mRNA AK029043.1 - 2 535
AK155160.1 - 2 571
BC030335.1 - 2 512 5401034

View Full Product Details