Gene Transcripts

Gene Symbol
Jazf1
Entrez Gene ID
231986
Gene Name
JAZF zinc finger 1
Gene Aliases
AI591476, C820002C15, Jaz1, Tip27
Chromosome Location
Chr. 6: 52768065 - 53068624 on Build GRCm38
UniGene ID
Mm.386865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168277.1 NP_001161749.1 5 1676
NM_173406.3 NP_775582.2 5 1675
GenBank mRNA AK047180.1 - 541
AK050474.1 - 5 1647

View Full Product Details