Gene Transcripts

Gene Symbol
Acad11
Entrez Gene ID
102632
Gene Name
acyl-Coenzyme A dehydrogenase family, member 11
Gene Aliases
5730439E10Rik, AI987948
Chromosome Location
Chr. 9: 104063703 - 104127646 on Build GRCm38
UniGene ID
Mm.269254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175324.3 NP_780533.2 20 3154
GenBank mRNA AK077531.1 - 20 3155
AK139918.1 - 20 3098
AK145561.1 - 20 3143
AK145834.1 - 20 3134
BC032994.1 - 13 2083

View Full Product Details