Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195632.1 | NP_001182561.1 | 12 | 1201 | |
NM_177379.4 | NP_796353.3 | 3 | 537 | ||
GenBank mRNA | AB088417.1 | - | 4 | 484 | |
AK036860.1 | - | 1 | 36 | ||
AY194286.1 | - | 3 | 264 | ||
BC132390.1 | - | 2 | 193 | 40130667 | |
BC138042.1 | - | 2 | 193 | 8861061 |