Gene Transcripts

Gene Symbol
Fam43a
Entrez Gene ID
224093
Gene Name
family with sequence similarity 43, member A
Gene Aliases
Tuf1
Chromosome Location
Chr. 16: 30599723 - 30602797 on Build GRCm38
UniGene ID
Mm.28244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177632.3 NP_808300.1 1 1201
GenBank mRNA AK083036.1 - 1 1201
AK137950.1 - 1 1202
BC099947.1 - 1 1214 30088416
BC138882.1 - 1 815 8861904
BC138884.1 - 1 791 8861906

View Full Product Details