Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177814.4 | NP_808482.2 | 16 | 3776 | |
GenBank mRNA | AK051416.1 | - | 6 | 1268 | |
AK122265.1 | - | 16 | 2566 | ||
AK147213.1 | - | 20 | 3929 | ||
BC056760.1 | - | 16 | 3776 | 6331997 |