Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177910.3 | NP_808578.1 | 2, 3 | 430 | |
GenBank mRNA | AK088295.1 | - | 2, 3 | 412 | |
AK148125.1 | - | 430 | |||
AK158470.1 | - | 2, 3 | 430 | ||
AK165816.1 | - | 2, 3 | 432 | ||
BC061207.1 | - | 3, 4 | 304 | 30249330 |