Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WAKL5
- Entrez Gene ID
-
838185
- Gene Name
- wall associated kinase-like 5
- Gene Aliases
-
AT1G16160, T24D18.24, T24D18_24, wall associated kinase-like 5
- Chromosome Location
-
Chr.1: 5535766 - 5538501 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101483.2 |
NP_173067.2 |
2 - 3 |
1218 |
84 |
View Full Product Details