Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AHA10
- Entrez Gene ID
-
838297
- Gene Name
- autoinhibited H[+]-ATPase isoform 10
- Gene Aliases
-
AT1G17260, F20D23.4, F20D23_4, autoinhibited H(+)-ATPase isoform 10
- Chromosome Location
-
Chr.1: 5904058 - 5908898 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101587.2 |
NP_173169.2 |
18 - 19 |
2408 |
78 |
View Full Product Details