Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCG11
Entrez Gene ID
838363
Gene Name
white-brown complex-like protein
Gene Aliases
ARABIDOPSIS THALIANA WHITE-BROWN COMPLEX HOMOLOG PROTEIN 11, AT1G17840, ATP-binding cassette G11, ATWBC11, AtABCG11, COF1, CUTICULAR DEFECT AND ORGAN FUSION 1, DESPERADO, DSO, F2H15.7, F2H15_7, WBC11, white-brown complex homolog protein 11
Chromosome Location
Chr.1: 6142402 - 6146521 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101647.5 NP_173226.2 8 - 9 2115 71
GenBank mRNA AY080792.1 - 8 - 9 1894 71
AY150457.1 - 8 - 9 1647 71

View Full Product Details