Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT1G18070
Entrez Gene ID
838387
Gene Name
Translation elongation factor EF1A/initiation factor IF2gamma family protein
Gene Aliases
AT1G18070, T10F20.8
Chromosome Location
Chr.1: 6213651 - 6218384 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035986.1 NP_001031063.1 12 - 13 963 117
NM_001198097.1 NP_001185026.1 12 - 13 968 117
NM_101669.3 NP_173247.1 12 - 13 1012 117
GenBank mRNA AY120769.1 - 12 - 13 947 117
BT008889.1 - 11 - 12 839 117

View Full Product Details