Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATLP-1
Entrez Gene ID
838405
Gene Name
Pathogenesis-related thaumatin superfamily protein
Gene Aliases
AT1G18250
Chromosome Location
Chr.1: 6276777 - 6278258 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035987.1 NP_001031064.1 2 - 3 533 76
NM_101683.2 NP_173261.1 2 - 3 638 76

View Full Product Details