Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HIK
- Entrez Gene ID
-
838418
- Gene Name
- ATP binding microtubule motor family protein
- Gene Aliases
-
ARABIDOPSIS NPK1-ACTIVATING KINESIN 1, AT1G18370, ATNACK1, F15H18.12, F15H18_12, HINKEL, NACK1, NPK1-ACTIVATING KINESIN 1
- Chromosome Location
-
Chr.1: 6319450 - 6324378 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101695.5 |
NP_173273.2 |
14 - 15 |
2858 |
90 |
GenBank mRNA |
AB081599.1 |
- |
13 - 14 |
2591 |
90 |
View Full Product Details