Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD30
Entrez Gene ID
838465
Gene Name
IQ-domain 30
Gene Aliases
AT1G18840, F6A14.7, F6A14_7, IQ-domain 30
Chromosome Location
Chr.1: 6500803 - 6504567 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035990.2 NP_001031067.1 7 - 8 2113 122
NM_001332375.1 NP_001322630.1 6 - 7 2231 122
NM_101741.4 NP_173318.2 7 - 8 2268 122
GenBank mRNA BT022035.1 - 7 - 8 1800 122

View Full Product Details