Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLH1
Entrez Gene ID
838554
Gene Name
chlorophyllase 1
Gene Aliases
AT1G19670, ATCLH1, ATHCOR1, CHLOROPHYLLASE, CORI1, CORONATINE-INDUCED PROTEIN 1, F6F9.28, F6F9_28, chlorophyllase 1
Chromosome Location
Chr.1: 6803486 - 6805092 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101823.4 NP_564094.1 1 - 2 444 81
GenBank mRNA AF021244.1 - 1 - 2 323 81
AY099815.1 - 1 - 2 322 81
BT000309.1 - 1 - 2 275 81

View Full Product Details