Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd32
Entrez Gene ID
838575
Gene Name
IQ-domain 32
Gene Aliases
AT1G19870, F6F9.8, F6F9_8, IQ-domain 32, IQD32
Chromosome Location
Chr.1: 6895009 - 6898818 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332417.1 NP_001320832.1 4 - 5 2956 66
NM_101842.5 NP_564097.1 5 - 6 2647 66

View Full Product Details