Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COR47
Entrez Gene ID
838632
Gene Name
cold-regulated 47
Gene Aliases
AT1G20440, AtCOR47, F5M15.22, F5M15_22, RD17, cold-regulated 47
Chromosome Location
Chr.1: 7084409 - 7086873 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101894.4 NP_173468.1 1 - 2 1548 130
GenBank mRNA AK230360.1 - - 1830 130
AY072392.1 - 1 - 2 445 130
AY114699.1 - 1 - 2 339 130

View Full Product Details