Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAK5
Entrez Gene ID
838718
Gene Name
wall associated kinase 5
Gene Aliases
AT1G21230, F16F4.9, F16F4_9, wall associated kinase 5
Chromosome Location
Chr.1: 7429895 - 7432477 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101976.2 NP_173546.1 2 - 3 1100 140

View Full Product Details