Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EXT3
- Entrez Gene ID
-
838728
- Gene Name
- extensin 3
- Gene Aliases
-
AT1G21310, ATEXT3, F16F4.4, F16F4_4, ROOT-SHOOT-HYPOCOTYL DEFECTIVE, RSH, extensin 3
- Chromosome Location
-
Chr.1: 7453245 - 7455009 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101983.4 |
NP_173553.1 |
1 - 2 |
1330 |
78 |
View Full Product Details